Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.08194

Sigma-Aldrich

HDAC6 Inhibitor III

Synonym(s):

HDAC6 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19N3O3
CAS Number:
Molecular Weight:
349.38
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

powder

potency

29 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

cream

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

General description

A cell-permeable, quinazolin-4-one based hydroxamic acid containing compound that acts as a potent and selective inhibitor of histone deacetylase 6 (HDAC6; IC50 = 29 nM). Exhibits high selectivity over other HDACs (IC50 = 1.88, 6.45, 1.75, and 4.08 µM for HDAC1, 2, 8, and 11, respectively). Does not affect hERG activity (>10 µM) or p450 activity (IC50 >6.5 µM). Blocks b-amyloid (Ab) aggregation in the presence of zinc (IC50 = 9.5 µM) and improves learning-based performance in mice. Induces neurite outgrowth (EC50 = 7.3 and 9.2 µM in PC12 and SH-SY5Y cells, respectively) and enhances synaptic activity (EC50 = 6.5 and 6.8 µM in PC12 and SH-SY5Y cells, respectively). Shown to enhance acetylated a-tubulin levels in hippocampus region of mice with Ab lesions.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Yu, C.W., et al. 2013. J. Med. Chem.56, 6775.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service