Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

506169

Sigma-Aldrich

InhibitorSelect p53 Pathway Regulators Panel

The InhibitorSelect p53 Pathway Regulators Panel controls the biological activity of InhibitorSelect. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

DMSO: soluble (supplied)

shipped in

wet ice

storage temp.

−20°C

General description

A panel containing 18 potent, selective and cell-permeable inhibitors and 1 activator that is useful for the study of p53 pathway:2 mg of ATM Kinase Inhibitor (Cat. No. 118500),5 mg ATM/ATR Kinase Inhibitor (Cat. No. 118501),10 mg of Anacardic Acid (Cat. No. 172050),5 mg of Cdk1 Inhibitor IV, RO-3306 (Cat. No. 217699),5 mg of Casein Kinase II Inhibitor III, TBCA (Cat. No. 218710),1 mg of Chk2 Inhibitor II (Cat. No. 220486),5 mg of Hdm2 E3 Ligase Inhibitor (Cat. No. 373225),5 mg of JNK Inhibitor II (Cat. No. 420119),1 mg of MDM2 Antagonist, Nutlin-3, Racemic (Cat. No. 444143),10 mg of MDMX Inhibitor, SJ-172550 (Cat. No. 444155),5 mg of MG-132 (Cat. No. 474790),5 mg of p21-Activated Kinase Inhibitor III, IPA-3 (Cat. No. 506106),5 mg of Pifithrin-α (Cat. No. 506132),1 mg of PI-103 (Cat. No. 528100),1 mg of PARP Inhibitor VIII, PJ34 (Cat. No. 528150),10 mg of PPM1D Phosphatase Inhibitor (Cat. No. 529578),10 mg of PRIMA-1 (Cat. No. 530050),1 mg of Caspase Inhibitor I (Cat. No. 627610),and 1 mg Trichostatin A, Streptomyces sp. (Cat. No. 647925).Also provided with 15 ml of anhydrous DMSO (Cat. No. KP31817). Supplied with a data sheet.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Each solutions is stable for up to 3 months at -20°C.
Please refer to information provided on the CD for reconstitution of individual inhibitors.

Other Notes

Feng, Z. and Levine, A.J., 2010. Trends Cell Biol.20, 427.
Wiman, K.G., 2010. Oncogene29, 4245.
Brown, C.J., et al. 2009. Nat. Rev. Cancer9, 862.
Dey, A., et al. 2008. Br. J. Cancer98, 4.
Levine, A.J., et al. 2006. Cell Death Differ.13, 1027.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service