Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.06075

Sigma-Aldrich

Novel Inducer of Death-1, NID-1

Synonym(s):

Novel Inducer of Death-1, NID-1, (E)-N-(2-ethylphenyl)-3-(5-nitrothiophen-2-yl)acrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2O3S
CAS Number:
Molecular Weight:
302.35
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, nitrothiophenylpropenamide compound that induces active, energy-dependent, and rapid non-apoptotic cell death (EC50 = 1.65 µM) in several transformed (DRD, RKO, H1299) and non-transformed (BJeH ) human cells, in a variety of cancer cell lines, and in mouse embryonic fibroblasts. The cell death appears to be non-necrotic, non-oxidative and is independent of DNA damage or cell proliferation. NID-1 induced cell death, although distinct from macroautophagy, does show PI 3-kinase suppression, ATG5-dependence, p62 degradation, and higher LC3 levels. Also, NID-1-induced death is independent of caspases, Bax and Bak and requires lysosomal activity and cytosolic vacuolization. Rescues mutant-huntingtin-induced cell death in a cell-based model of Huntington’s disease. Shown to increase cathepsin L levels, but does not affect cathepsins B, C, and H.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Varma, H., et al. 2013. Exp. Cell Res.319, 1759.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service