Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05983

Sigma-Aldrich

NS1738

Synonym(s):

NS1738, 1-(5-chloro-2-hydroxyphenyl)-3-(2-chloro-5-(trifluoromethyl)phenyl)urea, α7 nAChR positive allosteric modulator, NS1738, NS 1738, NS-1738

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl2F3N2O2
CAS Number:
Molecular Weight:
365.13
UNSPSC Code:
12352200

assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

InChI

1S/C14H9Cl2F3N2O2/c15-8-2-4-12(22)11(6-8)21-13(23)20-10-5-7(14(17,18)19)1-3-9(10)16/h1-6,22H,(H2,20,21,23)

InChI key

OUDXRNQPVSMGDW-UHFFFAOYSA-N

General description

A biarylurea derivative that acts as a selective positive allosteric modulator selective for α7 nicotinic acetylcholine receptors (α7 nAChR) without substantial activity for α4β2, α3β3 and α1-containing receptors. Exhibits cognitive enhancing effects in vivo.

Biochem/physiol Actions

Primary Target
α7 AChR

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Freitas, K., et al., 2013. J. Pharmacol. Exp. Ther.344, 264.
Timmermann, B., J. et al., 2007, Pharmacol. Exp. Ther.323, 294.
Faghih, R., 2007. Recent Pat CNS Drug Discov.2, 99.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service