Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05897

Sigma-Aldrich

(±)-PD 128907 Hydrochloride - CAS 112960-16-4 - Calbiochem

Synonym(s):

(±)-PD 128907 Hydrochloride - CAS 112960-16-4 - Calbiochem, Dopamine D₃ Receptor Agonist, PD 128,907, PD 128907, PD128907

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO3 · xHCl
CAS Number:
Molecular Weight:
249.31 (free base basis)
MDL number:
UNSPSC Code:
12352200

assay

≥98% (HPLC)

form

powder

potency

2.3 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 5 mg/mL

storage temp.

−20°C

General description

Selective dopamine D3 receptor agonist (Ki = 2.3 nM) with an 18-fold selectivity over long dopamine receptors (D2L) and exhibited only weak affinity for D4.2 receptors (Ki = 169 nM). Commonly used to reproduce cocaine′s discriminative stimulus effects and also in peripheral systemic studies of hypertension.

Biochem/physiol Actions

Primary Target
D₃

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Achat-Mendes, c. et al. 2010. J. Pharmacol. Exp. Ther.334, 556.

Zeng, C. et al. 2006. Circul. Res.99, 494.

Pugsley, T. A. et al. 1995. J. Pharmacol. Exp. Ther.275, 1355.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunyu Zeng et al.
Circulation research, 99(5), 494-500 (2006-08-12)
The dopaminergic and renin angiotensin systems interact to regulate blood pressure. Disruption of the D(3) dopamine receptor gene in mice produces renin-dependent hypertension. In rats, D(2)-like receptors reduce angiotensin II binding sites in renal proximal tubules (RPTs). Because the major
T A Pugsley et al.
The Journal of pharmacology and experimental therapeutics, 275(3), 1355-1366 (1995-12-01)
The present study determined the biochemical and pharmacological effects of PD 128907 [R-(+)-trans-3,4,4a,10b-tetrahydro-4-propyl-2H,5H- [1]benzopyrano[4,3-b]-1,4-oxazin-9-ol], a dopamine (DA) receptor agonist that shows a preference for the human D3 receptor. In transfected Chinese hamster ovary cells (CHO K1), PD 128907 displaced [3H]spiperone
Cindy Achat-Mendes et al.
The Journal of pharmacology and experimental therapeutics, 334(2), 556-565 (2010-05-25)
Dopamine (DA) D3 and D2 receptor mechanisms are implicated in cocaine's abuse-related behavioral effects, but the relative contribution of the two receptor subtypes is only partially characterized. This study investigated the role of D3 and D2 subtype mechanisms by determining

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service