Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05879

Sigma-Aldrich

L-741,626

Synonym(s):

L-741,626, Dopamine D₂ Receptor Antagonist, L-741,626, L-741626, L741626

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21ClN2O
CAS Number:
Molecular Weight:
340.85
MDL number:
UNSPSC Code:
12352200

assay

≥99% (HPLC)

Quality Level

form

solid

potency

7.1 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: soluble

storage temp.

2-8°C

InChI

1S/C20H21ClN2O/c21-17-7-5-16(6-8-17)20(24)9-11-23(12-10-20)14-15-13-22-19-4-2-1-3-18(15)19/h1-8,13,22,24H,9-12,14H2

InChI key

LLBLNMUONVVVPG-UHFFFAOYSA-N

General description

Selective dopamine D2 receptor antagonist that is shown to reversibly block agonist-induced activation of GIRK currents. Reported to have a 15-fold selectivity for D2R over D3R in vitro. Inhibition of 3H-spirperone binding to cloned rat Dopamine receptors show the following Ki values: D2R (7.1 nM), D3R (155 nM), D4R (596 nM). Commonly used D2 inhibitor in addiction research.

Biochem/physiol Actions

Primary Target
D₂

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Grundt, P. et al. 2007. Bioorg. Med. Chem. Lett.17, 745.

Caine, S. B. et al. 2002. J. Neurosci.22, 2977.
Pillai, G. et al. 1998. Neuropharmacol.337, 983.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service