Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05876

Sigma-Aldrich

RXFP1 Agonist

Synonym(s):

RXFP1 Agonist, 2-Isopropoxy-N-(2-(3-(trifluoromethylsulfonyl)phenylcarbamoyl)phenyl)benzamide, Leucine-Rich Repeat-Containing GPCR 7 Agonist, LGR7 Agonist, Relaxin/Insulin-like Family Peptide Receptor 1 Agonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21F3N2O5S
CAS Number:
Molecular Weight:
506.49
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable 2-acetamido-N-phenylbenzamide that selectively activates human, but not mouse, LGR7/RXFP1 (EC50 = 94 and 200 nM, respectively, in cAMP induction using HEK293-hRXFP1 transfectant and THP1) via allosteric interaction with the ECL3 (TM helix 5 to EC loop3; V646 to T660) region without competing against ECL2-mediated relaxin binding, while exhibiting cytotoxicity only at much higher concentrations (IC50 in 72 h = 9.4 µM in HEK293-RXFP1 cultures) and displaying no activity toward AVPR1B- (Arginine vasopressin receptor 1B) or LGR8/RXFP2-mediated cAMP induction. Although shown to be ~150-fold and 500-fold less potent than relaxin (RLX), respectively, in VEGF mRNA induction (275% of basal level in THP1 cells; 250 nM) and cellular impedance assays (in HEK293-RXFP1 cultures), pharmacokinetic studies reveal superior in vivo stability (T1/2 = 8.56 and 7.48 h in plasma and heart post single 30 mg/kg i.p. dosage in mice) to RLX (initial T1/2<10 min in human; i.v.) and, while RLX is not orally active, this chemical agonist is demonstrated to be bioavailable in mice via oral (Cmax/Tmax = 604 nM/1 h and 1026 ng/g/1.5 h in plasma and heart, respectively; 30 mg/kg) or intraperitoneal (Cmax/Tmax = 9.29 µM/1 h and 28.6 µmol/kg/1 h in plasma and heart, respectively; 30 mg/kg) administration in vivo with good aqueous solubility (7 µM in PBS).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
hRXFP1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Xiao, J., et al. 2013. Nat. Commun.4, 1953.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service