Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05483

Sigma-Aldrich

FFN511

Synonym(s):

FFN511, Fluorescent False Neurotransmitter 511, Vesicle Monoamine Transporter (VMAT2) Substrate, FFN511

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O2
CAS Number:
Molecular Weight:
284.35
UNSPSC Code:
12352200

assay

≥99% (HPLC)

Quality Level

form

solid

potency

1 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mM

storage temp.

−20°C

InChI

1S/C17H20N2O2/c18-6-5-11-10-15(20)21-17-13-4-2-8-19-7-1-3-12(16(13)19)9-14(11)17/h9-10H,1-8,18H2

InChI key

SKTIZSRPCXRDHX-UHFFFAOYSA-N

General description

A fluorescent false neurotransmitter that acts as an optical tracer that inhibits serotonin binding to VMAT2-containing membranes (IC50 = 1 µM). Compound has been used to assess the functional status of TH positive neurons by fluorescent measurement of uptatke and release.

Biochem/physiol Actions

Primary Target
VMAT2

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Dulcis, D. et al. 2013. Science340, 449.
Gubernator, N. G. et al. 2009. Science324, 1441.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niko G Gubernator et al.
Science (New York, N.Y.), 324(5933), 1441-1444 (2009-05-09)
The nervous system transmits signals between neurons via neurotransmitter release during synaptic vesicle fusion. In order to observe neurotransmitter uptake and release from individual presynaptic terminals directly, we designed fluorescent false neurotransmitters as substrates for the synaptic vesicle monoamine transporter.
Davide Dulcis et al.
Science (New York, N.Y.), 340(6131), 449-453 (2013-04-27)
Neurotransmitters have been thought to be fixed throughout life, but whether sensory stimuli alter behaviorally relevant transmitter expression in the mature brain is unknown. We found that populations of interneurons in the adult rat hypothalamus switched between dopamine and somatostatin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service