Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05481

Sigma-Aldrich

MRS 2211 - Calbiochem

Synonym(s):

MRS 2211 - Calbiochem, 2-[(2-Chloro-5-nitrophenyl)azo]-5-hydroxy-6-methyl-3-[(phosphonooxy)methyl]-4-pyridinecarboxaldehyde sodium salt, P2Y13 Purinergic Receptor Antagonist, MRS2211, MRS 2211, MRS-2211

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9ClN4Na3O8P
Molecular Weight:
496.64
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

water: 70 mM

storage temp.

−20°C

SMILES string

CC1=C(C(=C(C(=N1)N=NC2=C(C=CC(=C2)[N+](=O)[O-])Cl)COP(=O)([O-])[O-])C=O)O.[Na+].[Na+]

General description

A highly selective and competitive antagonist of P2Y13 purinergic receptor (pIC50 = 5.97). Exhibits about 20-fold greater selectivity over P2Y1 and P2Y12 receptors.

Biochem/physiol Actions

Primary Target
P2Y13

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kim, Y.C., et al. 2005. Biochem. Pharmacol.70, 266.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service