Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05146

Sigma-Aldrich

FMS/KIT Dual Kinase Inhibitor, PLX647

Synonym(s):

FMS/KIT Dual Kinase Inhibitor, PLX647, CDC2L6/CDK11/CDK19 Inhibitor, CDK8 Inhibitor, cFMS Inhibitor V, CSF-1 Receptor Inhibitor V, CSF1R Inhibitor V, DDR2 Inhibitor II, Flt-3 Inhibitor VIII, IL-34 Receptor Inhibitor V, NTRK3/TrkC Inhibitor, VEGFR Tyrosine Kinase Inhibitor XXXVI, VEGFR2 Tyrosine Kinase Inhibitor XXXIV, PLX-647, c-Kit Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17F3N4
CAS Number:
Molecular Weight:
382.38
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell-permeable and bio-available azaindolylmethylpyridamine derivative that acts as a potent inhibitor against FMS and KIT (IC50 = 28 and 16 nM, respectively) by targeting and locking these two PDGF family receptor kinases in their inactive DGF-out conformation, while being less effective against Flt3, KDR/VEGFR2, CDC2L6/CDK11/CDK19, NTRK3/TrkC, DDR2, CDK8/cyclin C (IC50 = 91, 130, 350, 620, 700, and 710 nM, respectively) and exhibiting much reduced or little potency toward other kinases in a 400-kinase panel selectivity study (IC50 >1 µM). Shown to effectively inhibit CSF-1-induced (with RANKL as co-stimulator) osteoclast differentiation (IC50 = 170 nM), cellular FLT3-ITD, FMS, KIT phosphorylations (IC50 ranges from 20 to 250 nM), as well as FLT3-ITD-, FMS-, KIT-dependent proliferations (IC50 ranges from 92 to 380 nM), while being less effective against KDR- or wt FLT3-dependent proliferations (IC50 ranges from 1.6 to 5 µM). Orally available in rats (AUC0-∞ (h·µg/mL)/Cmax (µg/mL) = 6.4/0.63 and 46.35/3.72 post 10 mg/kg and 40 mg/kg oral dosage, respectively) and mice (AUC0-∞ (h·µg/mL)/Cmax (µg/mL) = 8.51/1.67 and 53.3/7.78 post 10 mg/kg and 40 mg/kg oral dosage, respectively) and is reported to exhibit in vivo efficacy in preventing rat MRMT-1 breast carcinoma bone metastases-caused osteolysis (30 mg/kg b.i.d.) as well as in blocking inflammation responses in several murine models, including UUO- (unilateral ureter obstruction) induced kidney inflammation (40 mg/kg b.i.d.), collagen-induced arthritis (20 to 80 mg/kg b.i.d.), LPS-induced serum IL-6 & TNF-α release (single 40 mg/kg oral dose 4.25 h before LPS), and mast cell activation-induced anaphylaxis (single 80 mg/kg oral dose 2 h before antigen stimulation).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
FMS
Reversible: yes
Secondary Target
KIT

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, C., et al. 2013. Proc. Natl. Acad. Sci. USA110, 5689.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service