Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.04981

Sigma-Aldrich

Tie2/Trk Dual Inhibitor

Synonym(s):

Tie2/Trk Dual Inhibitor, Tie2 Inhibitor IV, Trk Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21ClN6O2
CAS Number:
Molecular Weight:
448.90
UNSPSC Code:
12352200

assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyrrolopyrimidinyl-phenyl urea that acts as a potent, reversible, ATP-competitive inhibitor against Tie2 and TrkA kinase activity in cell-free assays (IC50 = 40 and 0.4 nM, respectively; [ATP] = 100 µM) as well as Tie2- and TrkA-dependent tyrosine phosphorylation of cellular proteins upon ligand stimulation in cultures (IC50 = 3.7 and 4 nM, respectively, using respective NIH 3T3 transfectant; 1 h drug preincubation), displaying much reduced potency against PDGFR (IC50 =1.4 µM) and little or no inhibitory activity toward EGFR, FGFR, or KDR (IC50 >10 µM). Reported to be orally available in rat (systemic Cmax = 1,715 ng/mL; Tmax = 0.75 h; 4.2 mg/kg p.o. using ultrafasted rats) and efficacious in suppressing rat C6 glioblastoma tumor expansion (by 40%, 60%, and 70% in 10 d, respectively, with 10 mg/kg/d, 33 mg/kg/d, or 10 mg/kg/12 h oral dosing schedule) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TrkA
Reversible: yes
Secondary Target
Tie-2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Arcari, J.T., et al. 2013. Bioorg. Med. Chem. Lett.23, 3059.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service