Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.00486

Sigma-Aldrich

CXCR3 Antagonist

Synonym(s):

CXCR3 Antagonist, GPR9 Antagonist, 4-cyano-N-(3-fluoro-4-(1H-tetrazol-5-yl)benzyl)-N-(2-fluorobenzyl)benzenesulfonamide, AS612568, EMD1205395, CD183 Antagonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16F2N6O2S
CAS Number:
Molecular Weight:
466.46
UNSPSC Code:
12352200
Product 5.00486 is not currently sold in your country. Contact Technical Service

assay

≥95% (HPLC)

Quality Level

form

solid

potency

192 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

An arylsulfonamide derivative that acts as a CXCR3 antagonist (IC50 = 192 nM against CXCL10-induced chemotaxis of hCXCR3-overexpressing L1.2 cells) with good aqueous solubility (>100 µM at pH 7.4). Shown to be orally available in mice (83% bioavailability; t1/2 = 1 h; 10 mg/kg p.o.) in vivo and display a medium intrinsic clearance (CIint = 17 µL/min/mg) in in vitro mouse liver microsome stability test.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CXCR3
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C).

Other Notes

Crosignani, S., et al. 2010. Bioorg. Med. Chem. Lett.20, 3614.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service