Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

499250-M

Sigma-Aldrich

Osteonectin, Human Platelets

Synonym(s):

BM-40, SPARC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

Osteonectin purified from human platelets. Acidic, Ca2+-binding glycoprotein found in a variety of embryonic and adult tissues containing actively proliferating and remodeling cells. Differs from the bone-derived osteonectin in its glycosylation pattern and its collagen binding specificity.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 20 mM Tris-HCl, pH 7.4.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Kelm, R.J., et al. 1994. J. Biol. Chem.269, 30147.
Kelm, R.J. and Mann, K.G. 1991. J. Biol. Chem.266, 9632.
Sage, E.H. and Bornstein, P.J. 1991. J. Biol. Chem.266, 14831.
Villarreal, X.C., et al. 1989. Biochemistry28, 6483.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service