Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

496150

Sigma-Aldrich

Omi/HtrA2 Protease Inhibitor, Ucf-101

The Omi/HtrA2 Protease Inhibitor, Ucf-101, also referenced under CAS 313649-08-0, controls the biological activity of Omi/HtrA2 Protease. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Omi/HtrA2 Protease Inhibitor, Ucf-101, High Temperature Requirement A2 Inhibitor I, 5-[5-(2-Nitrophenyl)furfurylidine]-1,3-diphenyl-2-thiobarbituric Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H17N3O5S
CAS Number:
Molecular Weight:
495.51
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C27H19N3O5S/c31-25-22(17-20-15-16-24(35-20)21-13-7-8-14-23(21)30(33)34)26(32)29(19-11-5-2-6-12-19)27(36)28(25)18-9-3-1-4-10-18/h1-17,33-34H

InChI key

QMGXAGWZCHDZRG-UHFFFAOYSA-N

General description

A cell-permeable furfurylidine-thiobarbituric acid compound that acts as a potent, specific, competitive, and reversible inhibitor of the pro-apoptotic, heat-inducible, mitochondrial serine protease Omi/HtrA2 (IC50 = 9.5 µM for His-Omi134-458). Shows very little activity against various other serine proteases tested (IC50 ≥ 200 µM). Reported to block Omi/HtrA2 induced cell death in caspase-9 (-/-) null fibroblasts.
A cell-permeable, furfurylidine-thiobarbituric acid compound that acts as a potent, specific, competitive, and reversible inhibitor of the pro-apoptotic, heat-inducible, mitochondrial serine protease, Omi/HtrA2 (IC50 = 9.5 µM for His-Omi134-458). Reported to inhibit Omi/HtrA2-induced cell death in caspase-9 (-/-) null fibroblasts. Does not significantly inhibit other serine proteases, including kallikrein, trypsin, plasmin, thrombin, and tissue plasminogen activator (t-PA) (IC50 ≥200 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
pro-apoptotic, heat-inducible, mitochondrial serine protease Omi/HtrA2
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cilenti, L., et al. 2003. J. Biol. Chem.278, 11489.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service