Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4.91235

Sigma-Aldrich

Ethyldimethyl-(2-methoxyethyl)ammonium tris(pentafluoroethyl)trifluorophosphate

high purity

Synonym(s):

Ethyldimethyl-(2-methoxyethyl)ammonium tris(pentafluoroethyl)trifluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18F18NOP
CAS Number:
Molecular Weight:
577.23
UNSPSC Code:
12352107

assay

≥99.0% (electrophoresis)

form

liquid

storage temp.

2-30°C

General description

high purity

Analysis Note

Purity (electrophoresis): ≥ 99.0 %
Identity (NMR): passes test
Halides (IC): ≤ 100 ppm
Water (K. F.): ≤ 100 ppm

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service