Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4.91233

Sigma-Aldrich

1-(2-Methoxyethyl)-1-methylpyrrolidinium tris(pentafluoroethyl)trifluorophosphate

high purity

Synonym(s):

1-(2-Methoxyethyl)-1-methylpyrrolidinium tris(pentafluoroethyl)trifluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
589.24
UNSPSC Code:
12352107

Quality Level

form

liquid

General description

high purity

Application

1-(2-Methoxyethyl)-1-methylpyrrolidinium tris(pentafluoroethyl)trifluorophosphate high purity. CAS 1195983-48-2, molar mass 589.24 g/mol.

Analysis Note

Purity (electrophoresis)≥ 99.0 %Identity (NMR)passes testHalides (IC)≤ 100 ppmWater (K. F.)≤ 100 ppm

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service