Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4.90116

Sigma-Aldrich

1-Hexyl-3-methylimidazolium tetracyanoborate

for synthesis

Synonym(s):

1-Hexyl-3-methylimidazolium tetracyanoborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19BN6
CAS Number:
Molecular Weight:
282.15
UNSPSC Code:
12352107

assay

≥98.0% (HPLC)

form

liquid

pH

6 (20 °C in H2O, Aqueous solution)

mp

<-50 °C

density

0.99 g/cm3

storage temp.

2-30°C

Analysis Note

Purity (HPLC): ≥ 98.0 %
Identity (NMR-spectrum): passes test
Halides (IC): ≤ 0.1 %
Water (K. F.): ≤ 1.0 %

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service