Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4.90042

Sigma-Aldrich

N-Butylpyridinium chloride

for synthesis

Synonym(s):

N-Butylpyridinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14ClN
CAS Number:
Molecular Weight:
171.67
UNSPSC Code:
12352107

vapor pressure

0.000022 hPa ( 20 °C)

assay

≥98.0% (HPLC)

form

solid

pH

7 (20 °C in H2O)

mp

55-145 °C

storage temp.

2-30°C

InChI

1S/C9H14N/c1-2-3-7-10-8-5-4-6-9-10/h4-6,8-9H,2-3,7H2,1H3/q+1

InChI key

REACWASHYHDPSQ-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 98.0 %
Identity (NMR): passes test
Water (K. F.): ≤ 1.0 %

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service