Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4.90031

Sigma-Aldrich

1-Hexyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

for synthesis

Synonym(s):

1-Hexyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19F6N3O4S2
CAS Number:
Molecular Weight:
447.42
MDL number:
UNSPSC Code:
12352107
Product 4.90031 is not currently sold in your country. Contact Technical Service

assay

≥99.0% (HPLC)

form

liquid

pH

6 (20 °C in H2O)

mp

-9 °C

density

1.37 g/cm3

storage temp.

2-30°C

InChI

1S/C10H19N2.C2F6NO4S2/c1-3-4-5-6-7-12-9-8-11(2)10-12;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h8-10H,3-7H2,1-2H3;/q+1;-1

InChI key

RCNFOZUBFOFJKZ-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 99.0 %
Identity (NMR): passes test
Halides (IC): ≤ 0.01 %
Water (K. F.): ≤ 0.1 %

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service