Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

487600-M

Millipore

p-Nitrophenyl Phosphate, Disodium Salt, Tablets - CAS 4264-83-9 - Calbiochem

Each 175 mg tablet contains 5 mg of p-Nitrophenyl Phosphate, Disodium Salt plus an inert filler.

Synonym(s):

p-Nitrophenyl Phosphate, Disodium Salt, Tablets - CAS 4264-83-9 - Calbiochem, pNPP, 2Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4NO6P · 2Na · 6H2O
CAS Number:
Molecular Weight:
371.14
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

form

tablet

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

light yellow

storage temp.

2-8°C

InChI

1S/C6H6NO6P.2Na/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;;/h1-4H,(H2,10,11,12);;/q;2*+1/p-2

InChI key

VIYFPAMJCJLZKD-UHFFFAOYSA-L

General description

Each 175 mg tablet contains 5 mg of p-Nitrophenyl Phosphate, Disodium Salt (Cat. No. 4876) plus an inert filler. Dissolve one tablet in 5 ml of 10 mM diethanolamine, pH 9.5 containing 500 µM MgCl2, to prepare a 1 mg/ml solution.
Excellent substrate for alkaline phosphatase-based ELISA assays. Produces a yellow soluble end product that can be read at 405 nm to 410 nm. Both kinetic and endpoint determinations can be performed. Each tablet contains 5 mg of p-Nitrophenyl Phosphate, Disodium Salt (cat. no. 4876) plus an inert filler.
Note: 1 each = 100 tablets.

Biochem/physiol Actions

Primary Target
Substrate for phosphatase

Warning

Toxicity: Standard Handling (A)

Preparation Note

Dissolve one tablet in 5 ml of of 10 mM diethanolamine, pH 9.5 containing 0.5 mM MgCl₂ to prepare a 1 mg/ml stock solution.

Reconstitution

Unstable in solution; reconstitute just prior to use. Following reconstitution, aliquot and refrigerate (4°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service