Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

481487

Sigma-Aldrich

InhibitorSelect NF-κB Signaling Pathway Inhibitor Panel

The InhibitorSelect NF-κB Signaling Pathway Inhibitor Panel controls the biological activity of InhibitorSelect.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

DMSO: soluble (supplied)

shipped in

wet ice

storage temp.

−20°C

General description

A panel containing 14 potent and selective inhibitors useful for the study of NF-κB signaling pathway. This panel contains the following inhibitors: 10 mg of BAY 11-7082 (Cat. No. 196870); 1 mg of IKK Inhibitor VII (Cat. No. 401486); 5 mg of Interleukin1- Receptor-Associated-Kinase-1/4 Inhibitor (Cat. No. 407601); 10 mg of Histone Acetyltransferase Inhibitor II (Cat. No. 382110); 5 mg of MG-132 (Cat. No. 474790); 500 µg of NEMO-Binding Domain-Binding Peptide, Cell-Permeable (Cat. No. 480025); 500 µg of NF-κB SN50, Cell-Permeable Inhibitor Peptide (Cat. No. 481480); 5 mg of PD 98059 (Cat. No. 513000); 5 mg of PDK1/Akt/Flt Dual Pathway Inhibitor (Cat. No. 521275); 1 mg of Trichostatin A, Streptomyces sp. (Cat. No. 647925); 1 mg of TIRAP Inhibitor Peptide, Cell-Permeable (Cat. No. 613570), 1 mg of Tpl2 Kinase Inhibitor (Cat. No. 616373), 1 mg of (5Z)-7-Oxozeaenol, Curvularia sp. (Cat. No. 499610), and 5 mg of JNK Inhibitor II (Cat. No. 420119). Also provided with 15 ml of anhydrous DMSO.
A panel of 14 highly potent and selective inhibitors useful for the study of NF-κB signaling pathway. This panel contains the following inhibitors as well as 15 ml anhydrous DMSO for reconstitution:

Packaging

Packaged under inert gas

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) each inhibitor. Each inhibitor solution is stable for up to 3 months at -20°C.
Please refer to information provided on the CD for reconstitution information of individual inhibitors.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service