Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

480703-M

Millipore

Neurodazine

A cell-permeable trisubstituted imidazole compound that induces neurogenesis of non-pluripotent C2C12 myoblasts as well as cells derived from mature human skeletal muscle cultures (2 µM for 7-10 days).

Synonym(s):

Neurodazine, 2-(2-(5-(3-Chlorophenyl))furanyl)-4,5-bis-(4-methoxyphenyl)-imidazole, Nz

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H21ClN2O3
CAS Number:
Molecular Weight:
456.92
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

assay

≥98% (HPLC)

form

film

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

storage temp.

2-8°C

InChI

1S/C27H21ClN2O3/c1-31-21-10-6-17(7-11-21)25-26(18-8-12-22(32-2)13-9-18)30-27(29-25)24-15-14-23(33-24)19-4-3-5-20(28)16-19/h3-16H,1-2H3,(H,29,30)

InChI key

FEEOFPAEDSMOTO-UHFFFAOYSA-N

General description

A cell-permeable trisubstituted imidazole compound that induces neurogenesis of non-pluripotent C2C12 myoblasts as well as cells derived from mature human skeletal muscle cultures (2 µM for 7-10 days). Shown to preferentially upregulate genes involved in neurogenesis, but not those pertaining to myogenesis, in C2C12 myoblasts.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Williams, D.R., et al. 2007. J. Am. Chem. Soc.129, 9258.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

668.8 °F

flash_point_c

353.8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service