Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

480150-M

Millipore

Ras-Net (Elk-3) Pathway Inhibitor, XRP44X

The Ras-Net (Elk-3) Pathway Inhibitor, XRP44X, also referenced under CAS 729605-21-4, controls the biological activity of Ras-Net (Elk-3).

Synonym(s):

Ras-Net (Elk-3) Pathway Inhibitor, XRP44X, Actin Assembly Inhibitor XVI, Ras Activation Inhibitor, Tubulin Polymerization Inhibitor IX, 1-(3-Chlorophenyl)-4-((3-methyl-1-phenyl-1H-pyrazol-5-yl)carbonyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClN4O
CAS Number:
Molecular Weight:
380.87
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mM
ethanol: 50 mM

storage temp.

2-8°C

InChI

1S/C21H21ClN4O/c1-16-14-20(26(23-16)18-7-3-2-4-8-18)21(27)25-12-10-24(11-13-25)19-9-5-6-17(22)15-19/h2-9,14-15H,10-13H2,1H3

InChI key

NPHPNBGYPKBDDB-UHFFFAOYSA-N

General description

A cell-permeable pyrazole compound that is shown to prevent in vitro tubulin polymerization (IC50 = 0.2 µM) by targeting Colchicine (Cat. No. 234115) binding site (Kd = 0.3 µM) in cell-free assays and induce blebs formation in HUVEC cultures by perturbing both tubulin microtubles and actin cytoskeleton structures. Unrelated to its effects on actin and tubulin filaments, XRP44X is also reported to block the activations of Ras and its downstream effector molecules, such as Raf, MEK, Erk, Rsk, and Net (Elk-3/SAP-2/Erp), upon FGF-2 (Cat. No. 341595) stimulation. Potently inhibits the growth of HUVEC, NIH3T3, as well as 9 other cancer cell lines in antiproliferation assays (IC50 ≤11.3 nM).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wasylyk, C., et al. 2008. Cancer Res.68, 1275.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service