Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475858-M

Millipore

Met30 Antagonist, SMER3

The Met30 Antagonist, SMER3 controls the biological activity of Met30. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Met30 Antagonist, SMER3, Small-Molecule Enhancer of Rapamycin-3, 9H-Indeno[1,2-b][1,2,5]oxadiazolo[3,4-e]pyrazin-9-one, Indeno[1,2-b]furazano[3,4-E]pyrazin-9-one, SCF Met30 Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H4N4O2
Molecular Weight:
224.18
UNSPSC Code:
12352200
Pricing and availability is not currently available.

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

General description

A cell-permeable indenofurazinopyrazinone compound that is shown to directly target the F-box protein Met30 subunit of the Skp1-cullin-F-box (SCF) E3 ubiquitin ligase complex, disrupting Met30-Skp1 interaction and selectively inhibiting SCFMet30, but not SCFCdc4, E3 ubiquitin ligase activity. SMER3 is reported to act in a synthetic lethal fashion with rapamycin (Cat. Nos. 553210, 553211, & 553212) in both human lung cancer A549 and in yeast cultures. SMER3 treatment (10 to 30 µM) results in cell cycle arrest and morphological phenotype resembling that seen with temperature-sensitive Met30, but not Skp1 or Cdc4, mutant under non-permissive temperature in yeast cultures.

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
It is recommended to prepare a stock solution for 3 month use each time and store the unreconstituted material in solid form, protected from moisture and preferably under inert gas, for best long-term stability during storage.

Other Notes

Aghajan, M., et al. 2010. Nature Biotechnol.28, 738.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service