Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475816

Sigma-Aldrich

Microcystin-RR, Microcystis aeruginosa - CAS 111755-37-4 - Calbiochem

Synonym(s):

MCYST-RR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

assay

≥95% (HPLC)

form

solid

color

off-white

solubility

DMSO: 2.5 mg/mL
ethanol: soluble

General description

Arg-Arg analog of Microcystin-LR (Cat. No. 475815) that is less toxic. Inhibitor of protein-phosphatase 2A (IC50 = 3.4 nM). Useful for comparative structure and function studies.

Biochem/physiol Actions

Target IC50:3.4 nM inhibiting protein phosphatase 2A

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Not available for sale outside of the United States.

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Shirai et al.
Applied and environmental microbiology, 57(4), 1241-1245 (1991-04-01)
All samples of cyanobacterial blooms collected from 1986 to 1989 from Lake Kasumigaura, Ibaraki Prefecture, Japan, were hepatotoxic. The 50% lethal doses (LD50s) of the blooms to mice ranged from 76 to 556 mg/kg of body weight. Sixty-eight Microcystis cell
R Matsushima et al.
Biochemical and biophysical research communications, 171(2), 867-874 (1990-09-14)
Three microcystins, YR, LR and RR and nodularin, all of which are hepatotoxic compounds, inhibited dose-dependently the activity of protein phosphatase 2A in and the specific [3H]okadaic acid binding to a cytosolic fraction of mouse skin, as strongly as okadaic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service