Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

445825

Sigma-Aldrich

DL-2-Mercaptomethyl-3-guanidinoethylthiopropanoic Acid - Calbiochem

Synonym(s):

Plummer’s Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

assay

≥95% (HPLC)

Quality Level

form

solid

color

off-white

solubility

degassed water: soluble

General description

Potent reversible inhibitor of human plasma carboxypeptidase N (Ki = 2 nM).
Potent reversible inhibitor of human plasma carboxypeptidase N (Ki = 2 nM). Inhibits hydrolysis of bradykinin.

Biochem/physiol Actions

Target IC50:2 nM against human plasma carboxypeptidase N

Preparation Note

Addition of a small amount of NaOH or NH₄OH may be necessary to achieve complete solubilization.

Reconstitution

Unstable in solution; reconstitute just prior to use.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A potent mercapto bi-product analogue inhibitor for human carboxypeptidase N.
T H Plummer et al.
Biochemical and biophysical research communications, 98(2), 448-454 (1981-01-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service