Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444170-M

Millipore

Manumycin A, Streptomyces parvulus

An antibiotic with antitumor properties.

Synonym(s):

Manumycin A, Streptomyces parvulus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H38N2O7
CAS Number:
Molecular Weight:
550.64
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

assay

≥96% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow to brown

solubility

DMSO: 10 mg/mL
chloroform: soluble
methanol: soluble

storage temp.

2-8°C

InChI

1S/C31H38N2O7/c1-5-6-11-19(2)16-20(3)17-21(4)30(38)32-22-18-31(39,29-28(40-29)27(22)37)15-10-8-7-9-12-25(36)33-26-23(34)13-14-24(26)35/h7-10,12,15-19,28-29,34,39H,5-6,11,13-14H2,1-4H3,(H,32,38)(H,33,36)/b8-7+,12-9+,15-10+,20-16+,21-17+/t19-,28-,29-,31+/m1/s1

InChI key

TWWQHCKLTXDWBD-MVTGTTCWSA-N

General description

An antibiotic with antitumor properties. A cell-permeable potent and selective inhibitor of farnesyltransferase (FTase; IC50 = 5 µM) compared to geranylgeranyltransferase I (GGTase I; IC50 = 180 µM). Manumycin acts as a competitive inhibitor of FTase with respect to farnesylpyrophosphate (FPP; Ki = 1.2 µM), but acts as a noncompetitive inhibitor with respect to the Ras acceptor protein. Reported to inhibit the growth and invasive activity of pancreatic cancer cells.

Biochem/physiol Actions

Primary Target
Farnesyltransferase (FTAse)
Target IC50: 5 µM against farnesyltransferase (FTase); 180 µM against geranylgeranyltransferase I (GGTase I)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Kainuma, O., et al. 1997. Pancreas15, 379.
Nagase, T., et al. 1996. Int. J. Cancer 65, 620.
Hara, M., et al. 1995. Proc. Natl. Acad. Sci. USA92, 3333.
Hara, M., et al. 1993. Proc. Natl. Acad. Sci. USA90, 2281.
Tamanoi, R. 1993. Trends Biochem. Sci.18, 349.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service