Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

428035-M

Millipore

Anti-HOCl-Oxidized Low Density Lipoprotein Rabbit pAb

lyophilized, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

does not contain

preservative

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-HOCl-Oxidized Low Density Lipoprotein, rabbit polyclonal, recognizes HOCl-LDL. Weakly cross-reacts with HNE-LDL. It is validated for use in ELISA and IHC (frozen and paraffin sections).
Rabbit polyclonal antibody supplied as lyophilized undiluted serum. Recognizes HOCL-oxidized low density lipoprotein (LDL).
Recognizes HOCl-LDL. Weakly cross-reacts with HNE-LDL.

Immunogen

Human
human hypochlorite-modified low density lipoprotein (LDL)

Application

ELISA (1:400-1:4000)

Frozen Sections (<1:400)

Paraffin Sections (<1:400)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Reconstitute with 100 µl sterile distilled water. Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Does not cross-react with unmodified LDL. Weakly cross-reacts with HNE-LDL. Variables associated with assay conditions will dictate the optimal working dilution.
Lau, B.H. 2001. J. Nutrition131, 985s.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service