Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422720

Sigma-Aldrich

L-744,832

A cell-permeable potent and selective thiol-containing peptidomimetic farnesyltransferase (FTase) inhibitor with anti-tumor properties.

Synonym(s):

L-744,832, (2S)-2-[[(2S)-2-[(2S,3S)-2-[(2R)-2-Amino-3-mercaptopropyl]amino]-3-methylpentyl]oxy]-1-oxo-3-phenylpropyl]amino]-4-(methylsulfonyl)-butanoic Acid 1-Methylethyl Ester, L-744,382

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H45N3O6S2 · 2HCl
CAS Number:
Molecular Weight:
632.70
UNSPSC Code:
12352200

Quality Level

assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 15 mg/mL
DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C26H45N3O6S2/c1-6-19(4)23(28-15-21(27)17-36)16-34-24(14-20-10-8-7-9-11-20)25(30)29-22(12-13-37(5,32)33)26(31)35-18(2)3/h7-11,18-19,21-24,28,36H,6,12-17,27H2,1-5H3,(H,29,30)/t19-,21-,22+,23-,24+/m1/s1

InChI key

PGOKBMWPBDRDGN-ZXGKGEBGSA-N

General description

A cell-permeable potent and selective thiol-containing peptidomimetic farnesyltransferase (FTase) inhibitor with anti-tumor properties. Rapidly blocks p70S6K activation and DNA synthesis and promotes apoptosis in transgenic mice. Induces p21 expression and cell cycle arrest in the G1 phase. Displays synergistic effect with paclitaxel and epothilones in inhibiting tumor growth. Also mimics many of the effects of Rapamycin (Cat. No. 553210) and may be effective against tumors that exhibit inappropriate activation of the mTOR/p70S6K pathway.
A potent, cell-permeable, and selective thiol-containing peptidomimetic farnesyltransferase (FTase) inhibitor that blocks p70s6k activation and DNA synthesis and promote apoptosis in transgenic mice. Induces p21 expression and arrests cell in the G1 phase of the cell cycle. Also shown to act synergistically with paclitaxel and epothilones in inhibiting tumor growth and arresting cells in metaphase. Also mimics a number of the effects exhibited by Rapamycin (Cat. No. 553210) and may be effective against tumors that exhibit inappropirate activation of the mTOR/p70s6k pathway in animal models.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Other Notes

Law, B.K., et al. 2000. J. Biol. Chem.275, 10796.
Law, B.K., et al. 1999. J. Biol. Chem.274, 4743.
Barrington, R.E., et al. 1998. Mol. Cell. Biol.18, 85.
Moasser, M.M., et al. 1998. Proc. Natl. Acad. Sci. USA95, 1369.
Sepp-Lorenzino, L., and Rosen, N. 1998. J. Biol. Chem.273, 20243.
Kohl, N.E., et al. 1995. Nat. Med.1, 792.
Sepp-Lorenzino, L., et al. 1995. Cancer Res.55, 5302.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugènia Almacellas et al.
Autophagy, 17(3), 796-813 (2020-06-24)
Lysosomes, as primary degradative organelles, are the endpoint of different converging pathways, including macroautophagy. To date, lysosome degradative function has been mainly studied in interphase cells, while their role during mitosis remains controversial. Mitosis dictates the faithful transmission of genetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service