Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420131-M

Sigma-Aldrich

JNK Inhibitor III, Cell-Permeable, Negative Control

The JNK Inhibitor III, Cell-Permeable, Negative Control controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JNK Inhibitor III, Cell-Permeable, Negative Control, HIV-TAT₄₇₋₅₇-gaba-c-JUNδ₃₃₋₅₇, scrambled, Ac-YGRKKRRQRRR-gaba- DSNLIRGVLPTLMPKSLAQNKLKHA-NH₂, HIV-TAT₄₇₋₅₇-gaba-c-JUNδ₃₃₋₅₇, scrambled, Ac-YGRKKRRQRRR-gaba-DSNLIRGVLPTLMPKSLAQNKLKHA-NH₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C191H336N70O48S
Molecular Weight:
4413.22
UNSPSC Code:
12352200

Quality Level

assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 1 mg/mL

storage temp.

−20°C

General description

A cell-permeable 37 amino acid peptide that contains a scrambled sequence derived from the JNK binding domain (δ) of human c-Jun (amino acids 33-57) fused to the HIV-TAT transduction domain (amino acids 47-57) via a γ-aminobutyric acid (GABA) spacer. Does not disrupt c-Jun/JNK complex formation. Serves as a negative control for JNK Inhibitor III, Cell-Permeable (Cat. No. 420130).
A cell-permeable 37-mer peptide that contains a scrambled human c-Jun δ domain (amino acids 33-57) sequence linked to that of HIV-TAT protein transduction domain (amino acids 47-57) via a γ-aminobutyric acid (GABA) spacer. Does not disrupt c-Jun/JNK complex formation and thus serves as a negative control for JNK Inhibitor III, Cell-Permeable (Cat. No. 420130).

Biochem/physiol Actions

Primary Target
Serves as a negative control for JNK Inhibitor III, Cell-Permeable (Cat. No. 420130).

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-gaba-Asp-Ser-Asn-Leu-Ile-Arg-Gly-Val-Leu-Pro-Thr-Leu-Met-Pro-Lys-Ser-Leu-Ala-Gln-Asn-Lys-Leu-Lys-His-Ala-NH₂

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service