401478
IκB Kinase Inactive Control Peptide, Cell-Permeable
An inactive control for IκB Kinase Inhibitor Peptide.
Synonym(s):
IκB Kinase Inactive Control Peptide, Cell-Permeable, Ac- AAVALLPAVLLALLAPDDRHD AGLD AMKDE-NH₂
Select a Size
$278.00
Select a Size
About This Item
$278.00
Quality Level
assay
≥97% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
color
off-white to white
solubility
water: 1 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
Biochem/physiol Actions
An inactive control for IκB Kinase Inhibitor Peptide
Packaging
Warning
Sequence
Physical form
Reconstitution
Other Notes
Traenckner, E.B., et al. 1995. EMBO J.14, 2876.
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service