Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

401225

Sigma-Aldrich

Goat Anti-Mouse IgM, H&L Chain Specific Peroxidase Conjugate

Calbiochem®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

contains

0.02% Thimerosal as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

General description

Immunoaffinity purified goat polyclonal antibody conjugated to horseradish peroxidase. Recognizes mouse IgM, heavy and light chains.
This Goat Anti-Mouse IgM, H&L Chain Specific Peroxidase Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Mouse IgM, H&L Chain Specific.

Application

Enzyme Immunoassay (1:10,000)

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.

Other Notes

o-Phenylenediamine, 1 mg/ml in citrate buffer, pH 4.5, 0.1% urea peroxide at 25°C, was used as the substrate for testing immunoenzyme reactivity. Monospecific for mouse IgM, heavy and light chain, as determined by immunoelectrophoresis against normal mouse serum. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Lazar et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(17), 7672-7676 (1995-08-15)
We show that the higher plant Arabidopsis thaliana has a serine-arginine-rich (SR) protein family whose members contain a phosphoepitope shared by the animal SR family of splicing factors. In addition, we report the cloning and characterization of a cDNA encoding
María Rodríguez-Muñoz et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 32(4), 842-850 (2006-08-11)
The RGSZ1 and RGSZ2 proteins, members of the RGS-Rz subfamily of GTPase-activating proteins (GAP), are involved in Mu-opioid receptor desensitization. The expression of these proteins, as well as of their main target the Gz protein, is virtually restricted to the
Manoja K Brahma et al.
Journal of the American Heart Association, 9(15), e013039-e013039 (2020-08-05)
Background Perturbations in myocardial substrate utilization have been proposed to contribute to the pathogenesis of cardiac dysfunction in diabetic subjects. The failing heart in nondiabetics tends to decrease reliance on fatty acid and glucose oxidation, and increases reliance on ketone
Chae-Myeong Ha et al.
Journal of the American Heart Association, 12(19), e029898-e029898 (2023-09-26)
Background Lifestyle and metabolic diseases influence the severity and pathogenesis of cardiovascular disease through numerous mechanisms, including regulation via posttranslational modifications. A specific posttranslational modification, the addition of O-linked β-N acetylglucosamine (O-GlcNAcylation), has been implicated in molecular mechanisms of both
Heather M Berens et al.
Journal of virology, 85(8), 3858-3871 (2011-02-11)
Encephalitis induced by reovirus serotype 3 (T3) strains results from the apoptotic death of infected neurons. Extrinsic apoptotic signaling is activated in reovirus-infected neurons in vitro and in vivo, but the role of intrinsic apoptosis signaling during encephalitis is largely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service