Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4000-0061

Guava Express 7-AAD Reagent for Flow Cytometry

Cell impermeant nuclear dye used in viability assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NC.55

Quality Level

manufacturer/tradename

Guava®

compatibility

for use with EasyCyte Mini
for use with EasyCyte Plus
for use with EasyCyte
for use with PCA-96
for use with PCA

shipped in

wet ice

General description

細胞生存率アッセイに用いられる細胞不透過性色素です。すべての装置でご使用になれます。対応機種の詳細については、こちらの フローサイトメトリーの装置及びプラットフォーム をクリックしてください。

Legal Information

Guava is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service