370674
Guanosine 3′,5′-cyclic Monophosphorothioate, 8-Bromo-, Rp-Isomer, Sodium Salt
Synonym(s):
Guanosine 3′,5′-cyclic Monophosphorothioate, 8-Bromo-, Rp-Isomer, Sodium Salt, Rp-8-Br-cGMPS, Na, PKG Inhibitor II
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H10BrN5O6PS · Na
Molecular Weight:
462.15
UNSPSC Code:
12352200
assay
≥99% (HPLC)
Quality Level
form
lyophilized
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
solubility
water: 1 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
A potent cell-permeable, metabolically stable inhibitor of cGMP-dependent protein kinase (PKG). Significantly more lipophilic and membrane-permeable than cGMP or Rp-cGMPS. Resistant to mammalian cyclic nucleotide-dependent phosphodiesterases.
A potent, cell-permeable, and metabolically stable inhibitor of protein kinase G (PKG; Ki = 4 µM). Acts as a cyclic nucleotide-gated channel agonist (EC50 = 173.5 µM). Significantly more lipophilic and membrane-permeable than cGMP or Rp-cGMPS. Resistant to mammalian cyclic nucleotide-dependent phosphodiesterases. Note: 5 µmol = 2.31 mg.
Biochem/physiol Actions
Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target Ki: 4 µM against protein kinase G (PKG)
Warning
Toxicity: Standard Handling (A)
Other Notes
Wei, J.Y., et al. 1998. J. Mol. Neurosci. 10, 53.
Nakamura, A., et al. 1996. Br. J. Pharmacol.117, 407.
Van Uffelen, B.E., et al. 1996. J. Leukoc. Biol.60, 94.
Nakazawa, M., et al. 1994. Eur. J. Pharmacol.253, 179.
Zhuo, M., et al. 1994. Nature368, 635.
Nakamura, A., et al. 1996. Br. J. Pharmacol.117, 407.
Van Uffelen, B.E., et al. 1996. J. Leukoc. Biol.60, 94.
Nakazawa, M., et al. 1994. Eur. J. Pharmacol.253, 179.
Zhuo, M., et al. 1994. Nature368, 635.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service