Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345883

Sigma-Aldrich

GGTI-298 - CAS 180977-44-0 - Calbiochem

Synonym(s):

N-4-[2(R)-Amino-3-mercaptopropyl]amino-2-naphthylbenzoyl-(L)-Leucine methyl ester, TFA

Sign Into View Organizational & Contract Pricing


About This Item

assay

≥90% (HPLC)

Quality Level

form

solid

color

white

solubility

DMSO: 5 mg/mL

General description

A cell-permeable, prodrug form of the GGTase I inhibitor GGTI-297 (Cat. No. 345882). Inhibits the processing of Rap 1A (IC50 = 3 μM) but had no effect on the processing of H-Ras even at concentrations of 15 μM. Also arrests cells in the G0/G1 phase of the cell cycle, induces apoptosis and enhances nitric oxide synthase-2 induction by IL-1β.
A cell-permeable, prodrug form of the GGTase I inhibitor GGTI-297 (Cat. No. 345882). Inhibits the processing of Rap 1A (IC50 = 3 μM) but has no effect on the processing of H-Ras even at concentrations of 15 μM. Inhibits PDGF-receptor tyrosine phosphorylation. Also arrests cells in the G0/G1 phase of the cell cycle, induces apoptosis, and enhances iNOS induction by IL-1β.
A cell-permeable, prodrug form of the GGTase I inhibitor GGTI-297.

Biochem/physiol Actions

Target IC50:3 μM against processing of Rap 1A

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Degas DMSO just prior to reconstitution
Following reconstitution, aliquot and freeze (-20°C). Unstable in solution; reconstitute just prior to use.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Vogt et al.
The Journal of biological chemistry, 272(43), 27224-27229 (1997-10-27)
Recently we have shown that in fibroblasts (NIH 3T3 and Rat-1 cells) inhibition of protein geranylgeranylation leads to a G0/G1 arrest, whereas inhibition of protein farnesylation does not affect cell cycle distribution. Here we demonstrate that in human tumor cells
J D Finder et al.
The Journal of biological chemistry, 272(21), 13484-13488 (1997-05-23)
Recently, we have designed farnesyltransferase and geranylgeranyltransferase I inhibitors (FTI-277 and GGTI-298) that selectively block protein farnesylation and geranylgeranylation, respectively. In this study, we describe the opposing effects of these inhibitors on interleukin-1beta (IL-1beta)-stimulated induction of nitric-oxide synthase-2 (NOS-2) in
K Miquel et al.
Cancer research, 57(10), 1846-1850 (1997-05-15)
The mechanism by which the geranylgeranyltransferase I inhibitor GGTI-298 and the farnesyltransferase inhibitor FTI-277 inhibit human tumor growth is not known. Herein, we demonstrate that in the human lung adenocarcinoma A549 cells, GGTI-298 induced a G1-G0 block whereas FTI-277 induced
Y Qian et al.
Bioorganic & medicinal chemistry, 6(3), 293-299 (1998-05-06)
In this paper we describe the synthesis of a family of CAAL peptidomimetics as GGTase-I inhibitors. These inhibitors lack the central dipeptide AA in the key CAAL carboxy terminal sequence of geranylgeranylated proteins and are more selective for GGTase-I over
T F McGuire et al.
The Journal of biological chemistry, 271(44), 27402-27407 (1996-11-01)
We have used specific inhibitors for farnesyltransferase (FTase) and geranylgeranyltransferase (GGTase) I as well as combinations of lovastatin with geranylgeraniol (GGOH) or farnesol (FOH) to investigate the role of protein prenylation in platelet-derived growth factor (PDGF)-induced PDGF receptor tyrosine phosphorylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service