341598
Fibroblast Growth Factor-2 Inhibitor, NSC37204 - Calbiochem
The Fibroblast Growth Factor-2 Inhibitor, NSC37204 controls the biological activity of Fibroblast Growth Factor-2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Synonym(s):
Fibroblast Growth Factor-2 Inhibitor, NSC37204 - Calbiochem
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H14N2Na2O9S2
Molecular Weight:
548.45
UNSPSC Code:
12352200
assay
>95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
brown
solubility
DMSO: 5 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
A small molecule, compound that is shown to compete with the binding domain of TSP-1 against angiogenic factor FGF-2 (IC50 = 1.3 µM) in a dose-dependent manner and prevents the binding of FGF-2 to HSPG on endothelial cells (IC50 = 24.0 ± 6.9 µM), in vitro. It also demonstrates anti-growth properties in FGF-2-induced, but not serum-induced, endothelial cells (IC50 = 20.3 µM ± 5.9 µM). In CAM assays in vivo, where angiogenesis is induced by FGF-2 embedded in gelatin sponges, the presence of the small molecule (0.5 µg) reduces the mean number of blood vessels entering the sponge from 26 ±4 (FGF-2 alone) to 10 ± 2 (FGF-2 + NSC37203).
A small molecule, compound that is shown to compete with the binding domain of TSP-1 against angiogenic factor FGF-2 (IC50 = 1.3 µM) in a dose-dependent manner and prevents the binding of FGF-2 to HSPG on endothelial cells (IC50 = 24.0 ±6.9 µM), in vitro. It also demonstrates anti-growth properties in FGF-2-induced, but not serum-induced, endothelial cells (IC50 = 20.3 µM ±5.9 µM). In CAM assays in vivo, where angiogenesis is induced by FGF-2 embedded in gelatin sponges, the presence of the small molecule (0.5 µg) reduces the mean number of blood vessels entering the sponge from 26 ±4 (FGF-2 alone) to 10 ±2 (FGF-2 + NSC37203).
Warning
Toxicity: Standard Handling (A)
Other Notes
Colombo, G., et al. 2010. J. Biol. Chem.285, 8733.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service