341326
Fatty Acid Synthase Inhibitor, Cpd 10v - CAS 1027354-00-2 - Calbiochem
Synonym(s):
7-chloro-4-hydroxy-3-(2ʹ-methoxybiphenyl-3-yl)-2-oxo-1,2-dihydroquinoline-6-carbonitrile, FAS inhibitor
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
assay
≥98% (HPLC)
form
solid
color
white
solubility
DMSO: 50 mg/mL, clear, colorless
General description
A hydroxyquinolinone derivative that acts as a potent reversible inhibitor of type 1 fatty acid synthase (FAS) (IC50 = 19 nM) when enzyme activity is monitored by following the synthesis of free coenzyme A (CoA-SH).
The Fatty Acid Synthase Inhibitor, Cpd 10v, controls the biological activity of Fatty. This small molecule/inhibitor is primarily used for Cancer applications.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Alexey Rivkin et al.
Bioorganic & medicinal chemistry letters, 16(17), 4620-4623 (2006-06-21)
A series of 3-aryl-4-hydroxyquinolin-2(1H)-ones with fatty acid synthase inhibitory activity was prepared. Starting from a derivative with an IC(50) = 1.4 microM, SAR studies led to compounds with more than 70-fold increase in potency (IC(50) < 20 nM).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service