Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

328010

Sigma-Aldrich

ERK Inhibitor II, FR180204

InSolution, ≥97%

Synonym(s):

InSolution ERK Inhibitor II, FR180204

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13N7
Molecular Weight:
327.34
UNSPSC Code:
12352200

Quality Level

assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable pyrazolopyridazinamine that acts as a potent, ATP-competitive inhibitor of ERK1 and ERK2 (IC50 = 510 nM and 330 nM; Ki = 310 nM and 140 nM, respectively). Exhibits ~20-fold greater selectivity over p38α (IC50 = 10 µM) and shows little activity towards IKKα, MEK1, MKK4, PDGFRα, PKCα, Src, and Syk even at concentrations as high as 30 µM. Shown to inhibit TGF-β−induced AP-1 activation in Mv1Lu epithelial cells (IC50 of 3.1 µM). ERK Inhibitor II, Negative Control (Cat. No. 328008) and the solid form of this compound (Cat. No. 328007) are also available..
A cell-permeable pyrazolopyridazinamine that acts as a potent, ATP-competitive inhibitor of ERK1 and ERK2 (IC50 = 510 nM and 330 nM; Ki = 310 nM and 140 nM, respectively). Exhibits ~20-fold greater selectivity over p38α (IC50 = 10 µM) and shows little activity towards IKKα, MEK1, MKK4, PDGFRα, PKCα, Src, and Syk even at concentrations as high as 30 µM. Shown to inhibit TGF-β−induced AP-1 activation in Mv1Lu epithelial cells (IC50 of 3.1 µM). ERK Inhibitor II, Negative Control is also available (Cat. No. 328008).

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (500 µg/153 µl) solution of ERK Inhibitor II, FR180204 (Cat. No. 328007) in DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarit Cohen-Kedar et al.
European journal of immunology, 51(4), 864-878 (2021-02-23)
Intestinal epithelial cells (IECs) are the first to encounter luminal antigens and play an active role in intestinal immune responses. We previously reported that β-glucans, major fungal cell-wall glycans, induced chemokine secretion by IEC lines in a Dectin-1- and Syk-dependent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service