Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324878

Sigma-Aldrich

Focal Adhesion Kinase Inhibitor II

The Focal Adhesion Kinase Inhibitor II, also referenced under CAS 869288-64-2, controls the biological activity of Focal Adhesion Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Focal Adhesion Kinase Inhibitor II, PF-573,228, PF-228, 6-((4-((3-(Methanesulfonyl)benzyl)amino)-5-trifluoromethylpyrimidin-2-yl)amino)-3,4-dihydro-1H-quinolin-2-one, FAK Inhibitor II, Cdk7 Inhibitor V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20F3N5O3S
CAS Number:
Molecular Weight:
491.49
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C22H20F3N5O3S/c1-34(32,33)16-4-2-3-13(9-16)11-26-20-17(22(23,24)25)12-27-21(30-20)28-15-6-7-18-14(10-15)5-8-19(31)29-18/h2-4,6-7,9-10,12H,5,8,11H2,1H3,(H,29,31)(H2,26,27,28,30)

InChI key

HESLKTSGTIBHJU-UHFFFAOYSA-N

General description

A cell-permeable pyrimidinyldiamino compound that acts as a selective, ATP-competitive, and potent FAK inhibitor (IC50 = 4 nM). It inhibits CDK7/cycH/MAT1 and CDK1/cycB only at much higher concentrations (IC50 = 197 and 486 nM, respectively) and exhibits little or much reduced activity against 39 other commonly studied kinases (≤64% inhibition at 1 µM). PF-573,228 effectively inhibits cellular FAK Tyr397 phosphorylation in various human, rat, and canine cell lines (IC50 = ~30-500 nM during an 1 h incubation period) and concomitant blockage of Tyr31 phosphorylation of the FAK downstream substrate paxillin is also demonstrated in rat fibroblast REF52 cultures. A useful tool for studying FAK-mediated cellular functions.

Warning

Toxicity: Standard Handling (A)

Other Notes

Slack-Davis, J.K., et al. 2007. J. Biol. Chem.282, 14845.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chien-Chung Huang et al.
Cells, 10(8) (2021-08-28)
Angiogenesis is a critical process in the formation of new capillaries and a key participant in rheumatoid arthritis (RA) pathogenesis. Vascular endothelial growth factor (VEGF) stimulation of endothelial progenitor cells (EPCs) facilitates angiogenesis and the progression of RA. Phosphorylation of
Lohit Khera et al.
Life science alliance, 4(6) (2021-04-01)
Cancer stem cells (CSCs) are implicated in tumor initiation, metastasis and drug resistance, and considered as attractive targets for cancer therapy. Here we identified a clinically relevant signaling nexus mediated by AXL receptor, PYK2 and PKCα and show its impact

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service