Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324762

Sigma-Aldrich

Endopeptidase, Neutral, Porcine Kidney

Synonym(s):

CD10, Common Acute Lymphocytic Antigen, Endopeptidase-24.11, Enkephalinase, Microvillus Proteinase, Neprilysin

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202

assay

≥95% (HPLC)

form

lyophilized

specific activity

≥850 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

physiological buffer, pH 7.4: 1 mg/mL (e.g., Tris-buffered saline)

shipped in

ambient

storage temp.

−20°C

General description

Native, neutral endopeptidase from porcine kidney. A cell surface zinc-containing proteinase that is abundant in the brush border membranes of the kidney proximal tubules. Also present in the brain, endothelial cells, liver, and the lung tissue. Peptidase activity is specifically directed towards the cleavage on the amino side of hydrophobic amino acid residues. Contains an arginine in the active site that can interact with the C-terminal carboxylate of substrates.
Native, neutral endopeptidase from porcine kidney. A cell surface, zinc containing metallopeptidase that is abundant in the brush border membrane of the kidney proximal tubules. Also present in brain, endothelial cells, liver, and lung. Peptidase activity is specifically directed toward the cleavage on the amino side of hydrophobic residues. Contains an arginine in the active site that can interact with the C-terminal carboxylate of substrates.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit of enzyme will liberate 1.0 µmol pNA per min from Glutaryl-(Ala)₂-Phe-pNA at 25°C, pH 7.4.

Physical form

Lyophilized from 150 mM NaCl, 25 mM Tris-HCl, 5 mM CaCl₂, TRITON X-100 Detergent, pH 7.4.

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 3 days at 4°C.

Other Notes

Barnes, K., et al. 1995. J. Neurochem. 64, 1826.
Gafford, J.T., et al. 1983. Biochemistry 22, 3265.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service