Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324631

Millipore

Eg5 Inhibitor VII - CAS 912953-25-4 - Calbiochem

Synonym(s):

KSP Inhibitor VII; 6-(4-Trifluoromethyl)phenyl)-3,4-dihydro-2(1H)-quinolinone, GSK-1

Sign Into View Organizational & Contract Pricing


About This Item

assay

≥97% (HPLC)

form

powder

color

off-white

solubility

DMSO: 10 mg/mL

General description

A cell-permeable quinolinone compound that acts as a highly potent and selective Eg5/KSP inhibitor (Ki = 1.4, 2.1, 2.2, and 1.7 nM for the mutant A133D, KHC-L5, D130V, and the wild-type KSP, respectively) by antagonizing ATP binding via an allosteric site. Effectively induces monopolar spindles in ovarian adenocarcinoma SKOV3 cells and inhibits the proliferation of colorectal carcinoma HCT116 cells (IC50 = 36 and 0.5 nM againt the parental and the ispinesib-resistant D130V mutant line, respectively). It is uncompetitive with respect to microtubule and exhibits little activity against a panel of 7 other mitotic kinesins.
The Eg5 Inhibitor VII controls the biological activity of Eg5. This small molecule/inhibitor is primarily used for Cell Structure applications.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lusong Luo et al.
Nature chemical biology, 3(11), 722-726 (2007-10-09)
The mitotic kinesin KSP (kinesin spindle protein, or Eg5) has an essential role in centrosome separation and formation of the bipolar mitotic spindle. Its exclusive involvement in the mitotic spindle of proliferating cells presents an opportunity for developing new anticancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service