Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324378

Sigma-Aldrich

Domoic Acid, Mytilus edulis - CAS 14277-97-5 - Calbiochem

Synonym(s):

25-[2α, 3β, 4β (1Z, 3E, 5R)]-2-Carboxy-4-(5-carboxy-1-methyl-1,3-hexadienyl)-3-pyrrolidineacetic Acid

Sign Into View Organizational & Contract Pricing


About This Item

assay

≥95% (HPLC)

Quality Level

form

crystalline solid

color

white to off-white

solubility

methanol: 0.6 mg/mL
H2O: 8 mg/mL

General description

A heat-stable neurotoxin. Glutamate kainate receptor agonist that shows higher affinity for the kainate receptor than any other known kainic acid analog. Causes excessive excitation of neurons resulting in the depletion of energy stores, cellular edema, and death. Inhibits adenylate cyclase activity in neuronal membranes. Evokes Ca2+-dependent and Ca2+-independent release of GABA.
Glutamate kainate excitatory amino acid agonist with the highest affinity for the kainate receptor of all known kainic acid analogs. It has been used to purify the kainate receptor by affinity chromatography. Its binding to the receptor causes continuous stimulation of nerve cells leading to depletion of energy, cellular edema, and death. Its neurological effect resembles anterograde memory loss associated with Alzheimer′s disease. Inhibits adenylate cyclase activity in brain membranes. Evokes Ca2+-dependent and Ca2+-independent release of GABA.

Biochem/physiol Actions

Glutamate kainate receptor

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Do not freeze solutions. Stock solutions are stable for up to 24 h at 4°C.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Alfonso et al.
Neurochemistry international, 24(3), 267-274 (1994-03-01)
The effect of the neurotoxin domoic acid (DOM), a structural analogue of kainic acid, on the release of [3H]gamma-aminobutyric acid (GABA) and on the [Ca2+]i was studied in cultured chick retina cells. DOM stimulated dose-dependently the release of [3H]GABA with
G R Stewart et al.
Experimental neurology, 110(1), 127-138 (1990-10-01)
Domoic acid (Dom), a rigid analog of the excitotoxic amino acids, glutamate and kainic acid, is believed to be the mussel neurotoxin responsible for a recent food poisoning incident in Canada that killed some people and left others with memory
M S Nijjar et al.
Molecular and cellular biochemistry, 136(2), 105-111 (1994-07-27)
Adenylate cyclase activity measured by the formation of cyclic AMP in rat brain membranes was inhibited by a shellfish toxin, domoic acid (DOM). The inhibition of enzyme was dependent on DOM concentration, but about 50% of enzyme activity was resistant
P N Usherwood et al.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology, 103(1), 19-22 (1992-09-01)
1. A review is presented of recent advances in glutamate receptor research with particular emphasis on studies which show that some glutamate receptors in the central nervous systems (CNS) of Xenopus and rat contain a mixture of N-methyl-D-aspartate-sensitive and kainate-sensitive
R A Tasker et al.
Canadian journal of physiology and pharmacology, 69(3), 378-382 (1991-03-01)
Domoic acid, a structural analogue of kainic acid, has been identified as the toxin that poisoned people who consumed contaminated blue mussels harvested from eastern Prince Edward Island in December of 1987. To investigate the pharmacology of domoic acid in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service