Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

250600-M

Sigma-Aldrich

Cytochrome c, Equine Heart

Sign Into View Organizational & Contract Pricing


About This Item

assay

≥90% (A550)

form

lyophilized solid (salt-free)

solubility

H2O: 100 mg/mL

General description

Native cytochrome c purified from equine heart. Useful as a mitochondrial marker. Plays an important role in electron transport. Peripheral protein found on the outer face of the inner mitochodrial membrane. Supplied largely in the oxidized form.

Reconstitution

Following reconstitution, refrigerate (4 deg;C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 2 weeks at 4°, for up to 6 months at -20° C, and for up to 3 days at room temperature.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service