A cell-permeable antimicrobial that disrupts the CH1 domain structure of p300, an important mediator of tumor hypoxia adaptation. Prevents the association of p300 with CH1-interacting proteins, such as HIF-1α and STAT2. Shown to attenuate hypoxia-induced transcription in vitro (IC50 = 20 nM and <10 nM in inhibiting secreted and cellular VEGF, respectively, in Hep38 cells) and inhibit tumor growth by inducing necrosis in vivo (1 mg/kg, i.v., daily in tumor xenografted NCr/Nude mice). Exhibits no effect on the transcriptional activity of PAR, SREBP2, or SRC-1, whose association with p300 is not mediated via CH1 domain.
A cell-permeable dithiodiketopiperazine antimicrobial agent that disrupts the CH1 domain structure of p300.
Warning
Toxicity: Toxic (F)
Reconstitution
Following reconstitution in fresh, anhydrous DMSO, aliquot and freeze (-20°C). Stock solutions for up to 3 months at -20°C.
Other Notes
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Fujimoto, H., et al. 2004. J. Nat. Prod.67, 98. Kung, A.L., et al. 2004. Cancer Cell6, 33.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.