Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218841

Sigma-Aldrich

InSolution Caspase-9 Inhibitor I - Calbiochem

The InSolution Caspase-9 Inhibitor I controls the biological activity of Caspase-9. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

InSolution Caspase-9 Inhibitor I - Calbiochem, Z-LE(OMe)HD(OMe)-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H43FN6O10
Molecular Weight:
690.72
UNSPSC Code:
12352200

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A 5 mM (250 µg/72 µl) solution of Z-LEHD-FMK (Cat. No. 218761) in DMSO. A potent, cell-permeable, and irreversible inhibitor of caspase-9 (ICE-LAP6, Mch6).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-4, caspase-5, caspase-9
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Sequence

Z-Leu-Glu(OMe)-His-Asp(OMe)-CH₂F*

Physical form

A 5 mM solution of Z-LEHD-FMK (Cat. No. 218761) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

single spot by TLC

Other Notes

Fadeel, B., et al. 1999. Leukemia13, 719.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

188.6 °F - (refers to pure substance)

flash_point_c

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service