Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218712

Sigma-Aldrich

Anti-Casein Kinase II, β-Subunit Mouse mAb (6D5)

lyophilized, clone 6D5, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6D5, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

human, mouse, hamster, bovine

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

General description

Anti-Casein Kinase II, β-Subunit, mouse monoclonal, clone 6D5, recognizes recombinant CKII β-subunit and native holoenzyme. It is validated for Western blotting and immunocytochemistry.
Protein A purified mouse monoclonal antibody. Recognizes the ~25 kDa CKII β-subunit.
Recognizes recombinant CKII β-subunit and native holoenzyme.

Immunogen

Human
purified, recombinant, human CKII β-subunit

Application

Immunoblotting (5 µg/ml)

Immunocytochemistry (5 µg/ml)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from PBS.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Other Notes

Nastainczyk, W., et al. 1995. Hybridoma 14, 335.
Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aron Szabó et al.
PLoS biology, 11(8), e1001645-e1001645 (2013-09-10)
Phosphorylation is a pivotal regulatory mechanism for protein stability and activity in circadian clocks regardless of their evolutionary origin. It determines the speed and strength of molecular oscillations by acting on transcriptional activators and their repressors, which form negative feedback
D Vidal et al.
Plant biology (Stuttgart, Germany), 12(1), 134-144 (2010-07-27)
Two calcium- and light-dependent protein kinases have been reported in etiolated Cucumis sativus cotyledons (Vidal et al. 2007). In the present work, we studied casein kinase (CK) activity in etiolated cucumber cotyledons of in-gel and in vitro kinase assays, using
Tsuyoshi Oshima et al.
Science advances, 5(1), eaau9060-eaau9060 (2019-02-13)
Compounds targeting the circadian clock have been identified as potential treatments for clock-related diseases, including cancer. Our cell-based phenotypic screen revealed uncharacterized clock-modulating compounds. Through affinity-based target deconvolution, we identified GO289, which strongly lengthened circadian period, as a potent and
Janeen H Trembley et al.
Molecular and cellular biochemistry, 356(1-2), 21-35 (2011-07-16)
CK2 is a highly conserved, ubiquitous, signal responsive protein serine/threonine kinase. CK2 promotes cell proliferation and suppresses apoptosis, and increased CK2 expression is observed in all cancers examined. We previously reported that direct injection of antisense (AS) CK2α phosphorothioate oligonucleotides
Jiyeon Kim et al.
PloS one, 8(9), e74342-e74342 (2013-09-12)
The epithelial-to-mesenchymal transition (EMT) is a major phenotype of cancer metastasis and invasion. As a druggable cancer target, the inhibition of protein kinase CK2 (formally named to casein kinase 2) has been suggested as a promising therapeutic strategy to treat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service