Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

217605

Sigma-Aldrich

Anti-CD54 Mouse mAb (15.2)

liquid, ≥95% (SDS-PAGE), clone 15.2

Synonym(s):

Anti-ICAM-1, Domain 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$401.00

$401.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$401.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$401.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

15.2, monoclonal

assay

≥95% (SDS-PAGE)

form

liquid

contains

0.5 mg/mL gentamicin sulfate as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-CD54, mouse monoclonal, clone 15.2, recognizes D1 domain of the ~90 kDa CD54 (ICAM-1) in Raji cells. It is validated for FC, WB, IP, IHC on frozen sections, and neutralization studies.
Protein A purified mouse monoclonal antibody. Recognizes D1 domain of the ~90 kDa CD54 protein (ICAM-1).
Recognizes D1 domain of the ~90 kDa CD54 protein (ICAM-1) in Raji cells.

Immunogen

Human
human monocytes

Application

Flow Cytometry (1-10 µg/ml)

Frozen Sections (see comments)

Immunoblotting (see comments)

Immunoprecipitation (see comments)

Neutralization Studies (see comments)

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 100 mM KCl, 50 mM sodium phosphate, pH 7.5.

Reconstitution

Following initial use, aliquot and freeze (-20°C) for long-term storage. Avoid freeze/thaw cycles.

Analysis Note

Positive Control
Raji cells

Other Notes

FACS was performed on 5x105 cultured Raji human tumor cells using 1 µg/ml Anti-CD54 and FITC-conjugated goat anti-mouse IgG. This antibody will inhibit CD54 binding to LFA-1. This antibody has also been reported to work for frozen sections, immunoblotting, and immunoprecipitation. Variables associated with assay conditions will dictate the proper working dilution.
Reilly, P.L., et al. 1995. J. Immunol. 155, 529.
Berendt, A.R., et al. 1992. Cell68, 71.
Dransfield, I., et al. 1992. J. Cell Biol.116, 1527.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service