Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

208730

Sigma-Aldrich

Anti-Calpain-1/2, Small Subunit Mouse mAb (28F3)

liquid, clone 28F3, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

28F3, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, bovine

should not react with

mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

ambient

storage temp.

−20°C

General description

Purified mouse monoclonal antibody. Recognizes the ~28-30 kDa subunit of µ-calpain (calpain-1) and m-calpain (calpain-2).
Recognizes the ~28-30 kDa subunit of µ-calpain (calpain-1) and m-calpain (calpain-2). Recognizes both native and denatured forms of the small subunit of calpain.
This Anti-Calpain-1/2, Small Subunit Mouse mAb (28F3) is validated for use in ELISA, Immunoblotting, Immunoprecipitation for the detection of Calpain-1/2, Small Subunit.

Immunogen

Human Placenta
purified calpain from human placenta

Application

ELISA (1:2000)

Immunoblotting (1:2000)

Immunoprecipitation (1:500)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM sodium chloride, 20 mM sodium phosphate, 50% glycerol, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
MDBK cells

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaosi Li et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(47), 16807-16820 (2012-11-24)
Genetic factors strongly contribute to the pathogenesis of sporadic Alzheimer's disease (AD). Nevertheless, genome-wide association studies only yielded single nucleotide polymorphism loci of moderate importance. In contrast, microsatellite repeats are functionally less characterized structures within our genomes. Previous work has
Xiaopeng Wu et al.
Proteomics, 13(22), 3309-3326 (2013-10-12)
Cross-species transmissions of swine influenza viruses (SIVs) raise great public health concerns. In this study, subcellular proteomic profiles of human A549 cells inoculated with H3N2 subtype SIV were used to characterize dynamic cellular responses to infection. By 2DE and MS
Jarob Saker et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(3), 416-423 (2013-01-25)
Radioimmunotherapy is considered to have great potential for efficient and highly specific treatment of tumors. The aim of this study was to determine the efficacy of radioimmunotherapy when using (90)Y-labeled cetuximab and to determine to what degree induction and repair
Kerstin Seidel et al.
Brain pathology (Zurich, Switzerland), 21(1), 31-43 (2010-08-25)
Stroke is one of the major medical burdens in industrialized countries. Animal experiments indicate that blockade of the angiotensin AT1 receptor (AT1R) improves neurological outcome after cerebral ischemia. These protective effects are partially mediated by the angiotensin AT2 receptor (AT2R).
Lisa Gleißner et al.
Oncotarget, 8(67), 110983-110993 (2018-01-18)
The combination of radiotherapy and pharmacological inhibition of cellular signal transduction pathways offers promising strategies for enhanced cancer cell inactivation. However, the molecular effects of kinase inhibitors especially on DNA damage detection and repair after X-irradiation have to be understood

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service