Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

205615

Sigma-Aldrich

E-Cadherin Upregulator, ResEcad

A cell-permeable isoxazolocarboxamide compound that enhances the E-cadherin protein level in lung squamous cell carcinoma H520 and colorectal adenocarcinoma SW620 cells.

Synonym(s):

E-Cadherin Upregulator, ResEcad, 5-(Furan-2-yl)-N-(pyridine-4-yl)butyl)isoxazole-3-carboxamide, E-Cadherin Expression Restorer

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$135.00

$135.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$135.00

About This Item

Empirical Formula (Hill Notation):
C17H17N3O3
CAS Number:
Molecular Weight:
311.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

$135.00


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable isoxazolocarboxamide compound that enhances the E-cadherin protein level in lung squamous cell carcinoma H520 and colorectal adenocarcinoma SW620 cells (6.3- and 10-fold, respectively; 10 µM for 24 h) in a dose-dependent manner (EC50 = 1.25 and 2.13 µM, respectively), resulting in an effective culture invasiveness reduction (by ~50% in SW620 invasion assays). The observed cellular E-cadherin protein upregulation correlates well with an increase in both cellular E-cadherin gene CHD1 transcription (10.2-fold increase in SW620 by RNA-Seq analysis;10 µM for 24 h) and histone H4 Lys9 acetylation (19- and 57-fold enhancement in H529 and SW620 cells, respectively; 10 µM for 24 h), although evidence indicates no direct inhibition of HDACs 1-11 activity by the compound and other cellular factors involved in histone acetylation regulation are the likely candidates as the direct drug target(s).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Stoops, S.L., et al. 2011. ACS Chem. Biol.6, 452.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service