Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

205534

Millipore

C₁₂E₉, PROTEIN GRADE® Detergent, 10% Solution

A non-ionic detergent useful for the solubilization of membrane-bound proteins in their native state. HLB number: 13.6. Note: 1 set = 5 x 10 ml.

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

impurities

≤20 μM Peroxide

CMC

80 μM

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C14H30O2/c1-2-3-4-5-6-7-8-9-10-11-13-16-14-12-15/h15H,2-14H2,1H3

InChI key

SFNALCNOMXIBKG-UHFFFAOYSA-N

General description

A non-ionic detergent useful for the solubilization of membrane-bound proteins in their native state. CMC = 80 µM; HLB number = 13.6.
Note: 1 set = 5 x 10 ml.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Physical form

10% aqueous solution.

Other Notes

Megias, A., et al. 2001. Biochem. J.353, 521.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEIN GRADE is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

>300.2 °F

flash_point_c

> 149 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service